The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145315.1 | NP_001138787.1 | 2 - 3 | 216 | 105 |
NM_001145316.1 | NP_001138788.1 | 2 - 3 | 408 | 105 | |
NM_024918.3 | NP_079194.3 | 2 - 3 | 321 | 105 | |
XM_017028070.1 | XP_016883559.1 | 2 - 3 | 204 | 105 | |
XM_017028071.1 | XP_016883560.1 | 2 - 3 | 426 | 105 | |
GenBank mRNA | AK023408.1 | - | 2 - 3 | 187 | 105 |
AK093031.1 | - | 2 - 3 | 316 | 105 | |
BC058899.1 | - | 2 - 3 | 174 | 105 |