Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYM2
Entrez Gene ID
7750
Gene Name
zinc finger MYM-type containing 2
Gene Aliases
FIM, MYM, RAMP, SCLL, ZNF198
Chromosome Location
Chr.13: 19957412 - 20091845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190965.2 NP_001177894.1 2 - 3 196 63
NM_197968.3 NP_932072.1 2 - 3 242 63
XM_005266520.3 XP_005266577.1 2 - 3 201 63
XM_017020729.1 XP_016876218.1 2 - 3 107 63
XM_017020731.1 XP_016876220.1 2 - 3 185 63
XM_017020732.1 XP_016876221.1 2 - 3 207 63
XM_017020735.1 XP_016876224.1 2 - 3 184 63
XM_017020736.1 XP_016876225.1 2 - 3 356 63
XM_017020737.1 XP_016876226.1 2 - 3 131 63
XM_017020738.1 XP_016876227.1 2 - 3 185 63
GenBank mRNA AK226118.1 - 2 - 3 179 63
AK289741.1 - 2 - 3 167 63
AL136621.1 - 2 - 3 236 63
BC036372.1 - 2 - 3 242 63
BI460760.1 - 2 - 3 244 63
BX643074.1 - 2 - 3 191 63
BX647944.1 - 2 - 3 196 63

View Full Product Details