Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GREB1L
Entrez Gene ID
80000
Gene Name
growth regulation by estrogen in breast cancer 1 like
Gene Aliases
C18orf6, KIAA1772
Chromosome Location
Chr.18: 21242225 - 21526273 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142966.1 NP_001136438.1 17 - 18 2826 89
XM_006722547.2 XP_006722610.1 17 - 18 2789 89
XM_011526179.2 XP_011524481.1 17 - 18 2789 89
XM_017025988.1 XP_016881477.1 17 - 18 2789 89
XM_017025989.1 XP_016881478.1 17 - 18 2862 89
XM_017025990.1 XP_016881479.1 17 - 18 2789 89
XM_017025991.1 XP_016881480.1 17 - 18 2705 89
XM_017025992.1 XP_016881481.1 16 - 17 2660 89
XM_017025993.1 XP_016881482.1 16 - 17 2733 89
XM_017025994.1 XP_016881483.1 16 - 17 2462 89
XM_017025995.1 XP_016881484.1 16 - 17 2378 89
XM_017025996.1 XP_016881485.1 15 - 16 2333 89
GenBank mRNA AK023749.1 - 4 - 5 579 89
AK293321.1 - 5 - 6 773 89
BC125017.1 - 16 - 17 2384 89

View Full Product Details