The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142966.1 | NP_001136438.1 | 18 - 19 | 2961 | 88 |
XM_006722547.2 | XP_006722610.1 | 18 - 19 | 2924 | 88 | |
XM_011526179.2 | XP_011524481.1 | 18 - 19 | 2924 | 88 | |
XM_017025988.1 | XP_016881477.1 | 18 - 19 | 2924 | 88 | |
XM_017025989.1 | XP_016881478.1 | 18 - 19 | 2997 | 88 | |
XM_017025990.1 | XP_016881479.1 | 18 - 19 | 2924 | 88 | |
XM_017025991.1 | XP_016881480.1 | 18 - 19 | 2840 | 88 | |
XM_017025992.1 | XP_016881481.1 | 17 - 18 | 2795 | 88 | |
XM_017025993.1 | XP_016881482.1 | 17 - 18 | 2868 | 88 | |
XM_017025994.1 | XP_016881483.1 | 17 - 18 | 2597 | 88 | |
XM_017025995.1 | XP_016881484.1 | 17 - 18 | 2513 | 88 | |
XM_017025996.1 | XP_016881485.1 | 16 - 17 | 2468 | 88 | |
GenBank mRNA | AK023749.1 | - | 5 - 6 | 714 | 88 |
AK293321.1 | - | 6 - 7 | 908 | 88 | |
BC125017.1 | - | 17 - 18 | 2519 | 88 |