Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEK11
Entrez Gene ID
79858
Gene Name
NIMA related kinase 11
Gene Aliases
-
Chromosome Location
Chr.3: 131026850 - 131350465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146003.1 NP_001139475.1 12 - 13 1534 73
NM_001321220.1 NP_001308149.1 12 - 13 1534 73
NM_001321221.1 NP_001308150.1 14 - 15 1733 73
NM_001321222.1 NP_001308151.1 9 - 10 1219 73
NM_001321223.1 NP_001308152.1 12 - 13 1652 73
NM_024800.4 NP_079076.3 13 - 14 1607 73
NM_145910.3 NP_665917.1 13 - 14 1607 73
XM_011513167.2 XP_011511469.1 13 - 14 1807 73
XM_011513173.1 XP_011511475.1 11 - 12 1283 73
XM_011513174.1 XP_011511476.1 13 - 14 1553 73
XM_011513175.1 XP_011511477.1 12 - 13 1395 73
XM_011513181.1 XP_011511483.1 9 - 10 936 73
XM_017007205.1 XP_016862694.1 14 - 15 1910 73
XM_017007206.1 XP_016862695.1 14 - 15 1828 73
XM_017007207.1 XP_016862696.1 14 - 15 2044 73
XM_017007208.1 XP_016862697.1 13 - 14 1628 73
XM_017007209.1 XP_016862698.1 13 - 14 1837 73
XM_017007210.1 XP_016862699.1 13 - 14 1693 73
XM_017007211.1 XP_016862700.1 13 - 14 1628 73
XM_017007213.1 XP_016862702.1 13 - 14 1606 73
XM_017007214.1 XP_016862703.1 12 - 13 1709 73
XM_017007217.1 XP_016862706.1 10 - 11 1291 73
XM_017007218.1 XP_016862707.1 15 - 16 1811 73
XM_017007219.1 XP_016862708.1 12 - 13 1305 73
XM_017007220.1 XP_016862709.1 13 - 14 1628 73
XM_017007221.1 XP_016862710.1 13 - 14 1628 73
XM_017007222.1 XP_016862711.1 13 - 14 1628 73
XM_017007224.1 XP_016862713.1 14 - 15 1672 73
XM_017007225.1 XP_016862714.1 14 - 15 1685 73
XM_017007226.1 XP_016862715.1 14 - 15 1949 73
XM_017007227.1 XP_016862716.1 12 - 13 1429 73
XM_017007228.1 XP_016862717.1 10 - 11 1044 73
XM_017007229.1 XP_016862718.1 10 - 11 1291 73
XM_017007230.1 XP_016862719.1 10 - 11 1469 73
XM_017007232.1 XP_016862721.1 13 - 14 1612 73
XM_017007233.1 XP_016862722.1 12 - 13 1362 73
GenBank mRNA AB071996.1 - 13 - 14 1574 73
AB071997.1 - 13 - 14 1574 73
AK027148.1 - 8 - 9 800 73
AK297300.1 - 9 - 10 1219 73
AK316352.1 - 9 - 10 1134 73
AL833472.1 - 12 - 13 1505 73
BC028587.2 - 12 - 13 1651 73
BX328072.2 - 6 - 7 602 73

View Full Product Details