The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015395.2 | NP_056210.1 | 3 - 4 | 528 | 68 |
XM_005250253.3 | XP_005250310.1 | 3 - 4 | 500 | 68 | |
XM_017011937.1 | XP_016867426.1 | 3 - 4 | 512 | 68 | |
XM_017011938.1 | XP_016867427.1 | 3 - 4 | 512 | 68 | |
GenBank mRNA | AB037779.1 | - | 1 - 2 | 104 | 68 |
AK292996.1 | - | 3 - 4 | 430 | 68 | |
BC053591.1 | - | 3 - 4 | 433 | 68 |