Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr.7: 100400826 - 100429239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC002725.1 - 13 - 14 1544 129
KU178677.1 - 11 - 12 1106 129

View Full Product Details