Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM60
Entrez Gene ID
85025
Gene Name
transmembrane protein 60
Gene Aliases
C7orf35, DC32
Chromosome Location
Chr.7: 77793728 - 77798430 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032936.3 NP_116325.1 1 - 2 178 80
GenBank mRNA AF260336.1 - 1 - 2 179 80
BC065930.1 - 1 - 2 102 80
BX117806.1 - 1 - 2 178 80

View Full Product Details