Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMTM4
Entrez Gene ID
146223
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Gene Aliases
CKLFSF4
Chromosome Location
Chr.16: 66598178 - 66696707 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178818.2 NP_848933.1 5 - 5 3140 148
NM_181521.2 NP_852662.1 4 - 4 7843 148
XM_011522882.1 XP_011521184.1 4 - 4 7751 148
GenBank mRNA AF521889.1 - 5 - 5 3140 148
AK000855.1 - 1 - 1 1019 148
AL157468.1 - 1 - 1 1066 148
AW207043.1 - 1 - 1 148
CR933620.1 - 1 - 1 6752 148

View Full Product Details