Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTUD4
Entrez Gene ID
54726
Gene Name
OTU deubiquitinase 4
Gene Aliases
DUBA6, HIN1, HSHIN1
Chromosome Location
Chr.4: 145133650 - 145180019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005263079.4 XP_005263136.1 1 - 2 211 79
XM_011532040.2 XP_011530342.2 1 - 2 191 79
XM_011532041.2 XP_011530343.2 1 - 2 191 79

View Full Product Details