Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC29
Entrez Gene ID
26231
Gene Name
leucine rich repeat containing 29
Gene Aliases
FBL9, FBXL9
Chromosome Location
Chr.16: 67207139 - 67227062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004055.1 NP_001004055.1 3 - 4 407 59
NM_012163.2 NP_036295.1 3 - 4 572 59
XM_017023131.1 XP_016878620.1 3 - 4 419 59
XM_017023132.1 XP_016878621.1 3 - 4 722 59
XM_017023133.1 XP_016878622.1 3 - 4 748 59
XM_017023134.1 XP_016878623.1 3 - 4 527 59
GenBank mRNA AF176701.1 - 3 - 4 417 59
AK096101.1 - 1 - 2 1044 59
AK316602.1 - 3 - 4 420 59
BC018785.1 - 3 - 4 572 59

View Full Product Details