Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TANGO6
Entrez Gene ID
79613
Gene Name
transport and golgi organization 6 homolog
Gene Aliases
TMCO7
Chromosome Location
Chr.16: 68843547 - 69085182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024562.1 NP_078838.1 5 - 6 1141 107
XM_011523327.2 XP_011521629.1 5 - 6 1200 107
GenBank mRNA AB051533.1 - 5 - 6 1141 107
AK095333.1 - 4 - 5 687 107

View Full Product Details