The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020453.3 | NP_065186.3 | 7 - 8 | 1182 | 100 |
XM_005248119.4 | XP_005248176.1 | 7 - 8 | 1313 | 100 | |
XM_005248120.4 | XP_005248177.1 | 7 - 8 | 1300 | 100 | |
XM_011513722.2 | XP_011512024.1 | 6 - 7 | 1118 | 100 | |
XM_017008472.1 | XP_016863961.1 | 7 - 8 | 1293 | 100 | |
GenBank mRNA | AJ441078.1 | - | 7 - 8 | 1182 | 100 |
BC057839.1 | - | 7 - 8 | 1276 | 100 | |
BC131535.1 | - | 7 - 8 | 1160 | 100 |