Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LTV1
Entrez Gene ID
84946
Gene Name
LTV1 ribosome biogenesis factor
Gene Aliases
C6orf93, dJ468K18.4
Chromosome Location
Chr.6: 143843371 - 143863806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032860.3 NP_116249.2 6 - 7 905 78
GenBank mRNA AK027815.1 - 4 - 5 654 78
AK091447.1 - 1 - 2 3954 78
BC009855.2 - 6 - 7 905 78

View Full Product Details