The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032860.3 | NP_116249.2 | 8 - 9 | 1170 | 94 |
GenBank mRNA | AK027815.1 | - | 6 - 7 | 919 | 94 |
AK091447.1 | - | 3 - 4 | 4219 | 94 | |
BC009855.2 | - | 8 - 9 | 1170 | 94 |