Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPUSD4
Entrez Gene ID
84881
Gene Name
RNA pseudouridylate synthase domain containing 4
Gene Aliases
-
Chromosome Location
Chr.11: 126202094 - 126211692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144827.1 NP_001138299.1 1 - 2 242 69
NM_032795.2 NP_116184.2 1 - 2 242 69
XM_011543039.2 XP_011541341.1 1 - 2 212 69
GenBank mRNA AK027400.1 - 1 - 2 196 69
AK225320.1 - 1 - 2 202 69
AK308059.1 - 1 - 2 196 69
BC014131.2 - 1 - 2 203 69
DB051034.1 - 1 - 2 244 69

View Full Product Details