Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPUSD4
Entrez Gene ID
84881
Gene Name
RNA pseudouridylate synthase domain containing 4
Gene Aliases
-
Chromosome Location
Chr.11: 126202094 - 126211692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032795.2 NP_116184.2 4 - 5 704 86
XM_011543039.2 XP_011541341.1 4 - 5 701 86
GenBank mRNA AK027400.1 - 4 - 5 658 86
AK225320.1 - 4 - 5 664 86
AK300723.1 - 2 - 3 178 86
BC014131.2 - 4 - 5 665 86

View Full Product Details