Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ALG9
- Entrez Gene ID
-
79796
- Gene Name
- ALG9, alpha-1,2-mannosyltransferase
- Gene Aliases
-
CDG1L, DIBD1, GIKANIS, LOH11CR1J
- Chromosome Location
-
Chr.11: 111776096 - 111871581 on Build GRCh38
View Full Product Details