The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153256.3 | NP_694988.3 | 2 - 3 | 287 | 127 |
XM_011519438.2 | XP_011517740.1 | 3 - 4 | 606 | 127 | |
XM_017016028.1 | XP_016871517.1 | 2 - 3 | 1273 | 127 | |
GenBank mRNA | BC017269.2 | - | 2 - 3 | 287 | 127 |
BC048800.1 | - | 2 - 3 | 287 | 127 |