The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015059.2 | NP_055874.2 | 5 - 6 | 662 | 59 |
XM_005254708.4 | XP_005254765.1 | 8 - 9 | 1030 | 59 | |
XM_005254710.4 | XP_005254767.1 | 9 - 10 | 1168 | 59 | |
XM_005254711.4 | XP_005254768.1 | 8 - 9 | 1390 | 59 | |
XM_005254712.4 | XP_005254769.1 | 8 - 9 | 1047 | 59 | |
XM_005254713.4 | XP_005254770.1 | 8 - 9 | 1944 | 59 | |
XM_005254714.3 | XP_005254771.1 | 7 - 8 | 933 | 59 | |
XM_005254715.2 | XP_005254772.1 | 7 - 8 | 1639 | 59 | |
XM_006720717.3 | XP_006720780.1 | 8 - 9 | 1764 | 59 | |
XM_017022665.1 | XP_016878154.1 | 8 - 9 | 2115 | 59 | |
XM_017022666.1 | XP_016878155.1 | 8 - 9 | 974 | 59 | |
XM_017022667.1 | XP_016878156.1 | 6 - 7 | 836 | 59 | |
XM_017022668.1 | XP_016878157.1 | 8 - 9 | 1763 | 59 | |
XM_017022669.1 | XP_016878158.1 | 8 - 9 | 1764 | 59 | |
GenBank mRNA | AF402000.1 | - | 5 - 6 | 662 | 59 |