The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170585.1 | NP_001164056.1 | 36 - 37 | 2615 | 83 |
NM_153021.4 | NP_694566.4 | 37 - 38 | 2648 | 83 | |
XM_011532579.2 | XP_011530881.1 | 39 - 40 | 2955 | 83 | |
XM_011532581.2 | XP_011530883.1 | 39 - 40 | 2918 | 83 | |
XM_011532584.2 | XP_011530886.1 | 39 - 40 | 2955 | 83 | |
XM_011532588.2 | XP_011530890.1 | 39 - 40 | 2957 | 83 | |
XM_011532589.2 | XP_011530891.1 | 39 - 40 | 2952 | 83 | |
XM_011532590.2 | XP_011530892.1 | 39 - 40 | 2957 | 83 | |
XM_011532591.2 | XP_011530893.1 | 39 - 40 | 2954 | 83 | |
XM_011532593.2 | XP_011530895.1 | 35 - 36 | 2530 | 83 | |
XM_011532601.2 | XP_011530903.1 | 25 - 26 | 1943 | 83 | |
XM_011532602.2 | XP_011530904.1 | 24 - 25 | 1915 | 83 | |
XM_011532603.2 | XP_011530905.1 | 24 - 25 | 1992 | 83 | |
XM_011532609.2 | XP_011530911.1 | 22 - 23 | 1746 | 83 | |
XM_011532610.2 | XP_011530912.1 | 14 - 15 | 1932 | 83 | |
XM_017003432.1 | XP_016858921.1 | 38 - 39 | 2924 | 83 | |
XM_017003433.1 | XP_016858922.1 | 38 - 39 | 2890 | 83 | |
XM_017003434.1 | XP_016858923.1 | 39 - 40 | 2959 | 83 | |
GenBank mRNA | BC150633.1 | - | 37 - 38 | 2648 | 83 |
BC153864.1 | - | 36 - 37 | 2567 | 83 |