The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018088.2 | NP_001018098.1 | 56 - 57 | 7300 | 150 |
NM_017684.4 | NP_060154.3 | 54 - 55 | 7171 | 150 | |
NM_018080.3 | NP_060550.2 | 54 - 55 | 7171 | 150 | |
NM_020821.2 | NP_065872.1 | 56 - 57 | 7300 | 150 | |
XM_011521713.2 | XP_011520015.1 | 56 - 57 | 7261 | 150 | |
XM_011521714.2 | XP_011520016.1 | 56 - 57 | 7261 | 150 | |
GenBank mRNA | AJ608770.1 | - | 54 - 55 | 7154 | 150 |
AJ608771.1 | - | 56 - 57 | 7283 | 150 | |
AJ626860.1 | - | 54 - 55 | 7154 | 150 | |
AJ626861.1 | - | 56 - 57 | 7283 | 150 | |
AK128650.1 | - | 10 - 11 | 1714 | 150 | |
CR936716.1 | - | 6 - 6 | 781 | 219 |