The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139456.1 | NP_001132928.1 | 1 - 2 | 81 | 94 |
XM_005250143.3 | XP_005250200.1 | 2 - 3 | 485 | 94 |