The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139456.1 | NP_001132928.1 | 10 - 11 | 1067 | 73 |
NM_174959.3 | NP_777619.1 | 6 - 7 | 684 | 73 | |
XM_005250143.3 | XP_005250200.1 | 11 - 12 | 1471 | 73 | |
XM_011515797.2 | XP_011514099.1 | 7 - 8 | 758 | 73 | |
XM_017011746.1 | XP_016867235.1 | 8 - 9 | 873 | 73 | |
XM_017011747.1 | XP_016867236.1 | 7 - 8 | 698 | 73 | |
XM_017011748.1 | XP_016867237.1 | 7 - 8 | 703 | 73 | |
GenBank mRNA | BC036796.1 | - | 6 - 7 | 684 | 73 |
BX349158.2 | - | 1 - 2 | 114 | 73 | |
BX648789.1 | - | 6 - 7 | 4304 | 73 |