Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAT4
Entrez Gene ID
79633
Gene Name
FAT atypical cadherin 4
Gene Aliases
CDHF14, CDHR11, FAT-J, FATJ, HKLLS2, NBLA00548, VMLDS2
Chromosome Location
Chr.4: 125314913 - 125492932 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291285.1 NP_001278214.1 10 - 11 11904 79
NM_001291303.1 NP_001278232.1 10 - 11 11904 79
NM_024582.4 NP_078858.4 10 - 11 11898 79
XM_011532236.2 XP_011530538.1 11 - 12 12981 79
XM_011532237.2 XP_011530539.1 10 - 11 7789 79
GenBank mRNA AK096646.1 - 2 - 3 200 79
AK291461.1 - 2 - 3 444 79
KJ131486.1 - 2 - 3 141 79

View Full Product Details