The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178563.3 | NP_848658.3 | 12 - 13 | 2157 | 117 |
XM_017012136.1 | XP_016867625.1 | 12 - 13 | 3479 | 117 | |
XM_017012137.1 | XP_016867626.1 | 12 - 13 | 3761 | 117 | |
XM_017012138.1 | XP_016867627.1 | 11 - 12 | 1960 | 117 | |
XM_017012140.1 | XP_016867629.1 | 11 - 12 | 1963 | 117 | |
XM_017012141.1 | XP_016867630.1 | 11 - 12 | 3296 | 117 | |
XM_017012142.1 | XP_016867631.1 | 11 - 12 | 1963 | 117 | |
XM_017012143.1 | XP_016867632.1 | 11 - 12 | 1963 | 117 | |
GenBank mRNA | AK302827.1 | - | 12 - 13 | 2157 | 117 |
BC030651.2 | - | 13 - 14 | 2265 | 117 |