The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178563.3 | NP_848658.3 | 6 - 7 | 813 | 80 |
XM_011516122.2 | XP_011514424.1 | 5 - 6 | 619 | 80 | |
XM_017012136.1 | XP_016867625.1 | 6 - 7 | 2135 | 80 | |
XM_017012137.1 | XP_016867626.1 | 6 - 7 | 2417 | 80 | |
XM_017012138.1 | XP_016867627.1 | 5 - 6 | 619 | 80 | |
XM_017012139.1 | XP_016867628.1 | 5 - 6 | 619 | 80 | |
XM_017012140.1 | XP_016867629.1 | 5 - 6 | 619 | 80 | |
XM_017012141.1 | XP_016867630.1 | 5 - 6 | 1952 | 80 | |
XM_017012142.1 | XP_016867631.1 | 5 - 6 | 619 | 80 | |
XM_017012143.1 | XP_016867632.1 | 5 - 6 | 619 | 80 | |
GenBank mRNA | AK302827.1 | - | 6 - 7 | 813 | 80 |
BC030651.2 | - | 6 - 7 | 788 | 80 |