Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHGC5
Entrez Gene ID
56097
Gene Name
protocadherin gamma subfamily C, 5
Gene Aliases
PCDH-GAMMA-C5
Chromosome Location
Chr.5: 141489241 - 141512979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032407.1 NP_115783.1 1 - 1 2569 90
GenBank mRNA AF152526.1 - 1 - 1 2569 90
BC112081.1 - 1 - 1 2643 90
BC112083.1 - 1 - 1 2643 90

View Full Product Details