The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032011.1 | NP_114400.1 | 1 - 1 | 2415 | 110 |
GenBank mRNA | AF152510.1 | - | 1 - 1 | 2415 | 110 |