The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286823.1 | NP_001273752.1 | 3 - 4 | 311 | 147 |
NM_017820.4 | NP_060290.3 | 3 - 4 | 311 | 147 | |
XM_011518810.1 | XP_011517112.1 | 3 - 4 | 311 | 147 | |
XM_017014867.1 | XP_016870356.1 | 3 - 4 | 306 | 147 | |
GenBank mRNA | AK094438.1 | - | 3 - 4 | 211 | 147 |
AL536907.3 | - | 3 - 4 | 182 | 147 | |
BC037355.1 | - | 3 - 4 | 213 | 147 | |
BC110879.1 | - | 3 - 4 | 219 | 147 | |
DA071548.1 | - | 3 - 4 | 211 | 147 | |
DA328221.1 | - | 3 - 4 | 311 | 147 |