The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182973.1 | NP_892018.1 | 7 - 8 | 1011 | 90 |
XM_011527978.2 | XP_011526280.1 | 9 - 10 | 1374 | 90 | |
GenBank mRNA | AB109390.1 | - | 8 - 9 | 1128 | 90 |
AJ488946.1 | - | 7 - 8 | 1011 | 90 | |
AJ488947.1 | - | 7 - 8 | 1011 | 90 |