Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTN3
Entrez Gene ID
4917
Gene Name
netrin 3
Gene Aliases
NTN2L
Chromosome Location
Chr.16: 2471499 - 2474145 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006181.2 NP_006172.1 1 - 2 1133 86
GenBank mRNA U86759.1 - 1 - 2 1133 86

View Full Product Details