Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARID5A
Entrez Gene ID
10865
Gene Name
AT-rich interaction domain 5A
Gene Aliases
MRF-1, MRF1, RP11-363D14
Chromosome Location
Chr.2: 96536719 - 96552638 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319085.1 NP_001306014.1 2 - 3 230 119
NM_001319087.1 NP_001306016.1 2 - 3 230 119
NM_001319092.1 NP_001306021.1 2 - 3 561 119
NM_001319094.1 NP_001306023.1 2 - 3 230 119
NM_212481.2 NP_997646.1 2 - 3 230 119
XM_011510499.2 XP_011508801.1 2 - 3 2098 119
XM_017003183.1 XP_016858672.1 3 - 4 576 119
GenBank mRNA AK300173.1 - 2 - 3 197 119
AK304566.1 - 2 - 3 197 119
BC047390.1 - 2 - 3 564 119
BM556343.1 - 2 - 3 175 119
BX396215.2 - 2 - 3 180 119
CR981009.1 - 2 - 3 185 119
HY002073.1 - 2 - 3 230 119
M62324.1 - 2 - 3 182 119

View Full Product Details