The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286479.1 | NP_001273408.1 | 5 - 6 | 700 | 88 |
NM_001286480.1 | NP_001273409.1 | 6 - 7 | 822 | 88 | |
NM_014783.4 | NP_055598.1 | 5 - 6 | 1441 | 88 | |
NM_199357.1 | NP_955389.1 | 5 - 6 | 1441 | 88 | |
GenBank mRNA | AK303025.1 | - | 6 - 7 | 741 | 88 |
AK316437.1 | - | 5 - 6 | 700 | 88 | |
BC039563.2 | - | 5 - 6 | 1296 | 88 | |
BC050545.1 | - | 5 - 6 | 1396 | 88 | |
BC063444.1 | - | 5 - 6 | 1392 | 88 | |
D87717.2 | - | 5 - 6 | 1441 | 88 |