The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006420.2 | NP_006411.2 | 27 - 28 | 3911 | 66 |
XM_005260252.3 | XP_005260309.1 | 27 - 28 | 3925 | 66 | |
GenBank mRNA | AB209397.1 | - | 7 - 8 | 867 | 66 |
AF084521.1 | - | 27 - 28 | 3759 | 66 | |
AK022623.1 | - | 6 - 7 | 733 | 66 | |
AK074303.1 | - | 10 - 11 | 1287 | 66 | |
AK302526.1 | - | 13 - 14 | 1766 | 66 | |
BC144625.1 | - | 26 - 27 | 3941 | 66 | |
BC144626.1 | - | 28 - 29 | 4696 | 66 |