The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006420.2 | NP_006411.2 | 29 - 30 | 4200 | 98 |
XM_005260252.3 | XP_005260309.1 | 29 - 30 | 4214 | 98 | |
GenBank mRNA | AB209397.1 | - | 9 - 10 | 1156 | 98 |
AF084521.1 | - | 29 - 30 | 4048 | 98 | |
AK022623.1 | - | 8 - 9 | 1022 | 98 | |
AK074303.1 | - | 12 - 13 | 1576 | 98 | |
AK302526.1 | - | 15 - 16 | 2055 | 98 | |
BC144625.1 | - | 28 - 29 | 4230 | 98 | |
BC144626.1 | - | 30 - 31 | 4985 | 98 |