The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015965.6 | NP_057049.5 | 2 - 3 | 200 | 73 |
GenBank mRNA | AF261134.1 | - | 2 - 3 | 631 | 73 |
AF286697.1 | - | 2 - 3 | 178 | 73 | |
AK293859.1 | - | 2 - 3 | 182 | 73 | |
BC000589.2 | - | 2 - 3 | 179 | 73 | |
BC009189.2 | - | 2 - 3 | 187 | 73 | |
BG705222.1 | - | 2 - 3 | 189 | 73 | |
BM839920.1 | - | 2 - 3 | 200 | 73 | |
BP230223.1 | - | 2 - 3 | 180 | 73 |