The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003658.4 | NP_003649.2 | 2 - 3 | 538 | 89 |
XM_011543043.1 | XP_011541345.1 | 2 - 3 | 365 | 89 | |
XM_011543044.2 | XP_011541346.1 | 2 - 3 | 383 | 89 | |
GenBank mRNA | AF031924.1 | - | 2 - 3 | 582 | 89 |
AJ243512.1 | - | 2 - 3 | 504 | 89 | |
BC069378.1 | - | 2 - 3 | 538 | 89 | |
BC111432.1 | - | 2 - 3 | 507 | 89 | |
BC111572.1 | - | 2 - 3 | 507 | 89 |