Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU2AF1
Entrez Gene ID
5450
Gene Name
POU class 2 associating factor 1
Gene Aliases
BOB1, OBF-1, OBF1, OCAB
Chromosome Location
Chr.11: 111352251 - 111379432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006235.2 NP_006226.2 3 - 4 444 64
XM_005271593.1 XP_005271650.1 4 - 5 586 64
XM_005271594.3 XP_005271651.1 4 - 5 543 64
XM_006718859.1 XP_006718922.1 4 - 5 366 64
XM_006718860.3 XP_006718923.1 4 - 5 586 64
XM_017017932.1 XP_016873421.1 3 - 4 287 64
GenBank mRNA AK313573.1 - 3 - 4 287 64
BC032549.1 - 3 - 4 271 64
X83504.1 - 3 - 4 257 64
Z47550.1 - 3 - 4 285 64
Z49194.1 - 3 - 4 712 64

View Full Product Details