Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMAB
Entrez Gene ID
326625
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblB type
Gene Aliases
ATR, CFAP23, cblB, cob
Chromosome Location
Chr.12: 109553715 - 109573553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052845.3 NP_443077.1 8 - 9 720 95
NR_038118.1 9 - 10 807 95
XM_011538267.2 XP_011536569.1 8 - 9 837 95
XM_011538268.2 XP_011536570.1 6 - 7 1052 95
XM_011538269.2 XP_011536571.1 6 - 7 852 95
GenBank mRNA AK295211.1 - 9 - 10 828 95
AK312598.1 - 8 - 9 712 95
BC005054.1 - 8 - 9 660 95
BC011831.2 - 8 - 9 664 95

View Full Product Details