Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS37C
Entrez Gene ID
55048
Gene Name
VPS37C, ESCRT-I subunit
Gene Aliases
-
Chromosome Location
Chr.11: 61130256 - 61161444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017966.4 NP_060436.4 4 - 5 406 61
XM_005274077.3 XP_005274134.1 4 - 5 447 61
GenBank mRNA AK302384.1 - 4 - 5 474 61
AL834261.1 - 4 - 5 414 61
BC141827.1 - 4 - 5 369 61
BC142702.1 - 4 - 5 369 61
BM450171.1 - 3 - 4 376 61

View Full Product Details