The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000343.3 | NP_000334.1 | 3 - 4 | 563 | 60 |
NM_001256314.1 | NP_001243243.1 | 2 - 3 | 281 | 60 | |
XM_011530331.1 | XP_011528633.1 | 3 - 4 | 545 | 60 | |
GenBank mRNA | AK297665.1 | - | 2 - 3 | 255 | 60 |
AK312948.1 | - | 3 - 4 | 467 | 60 | |
AK316184.1 | - | 2 - 3 | 260 | 60 | |
BC139839.1 | - | 3 - 4 | 332 | 60 | |
CR456579.1 | - | 3 - 4 | 363 | 60 | |
M24847.1 | - | 3 - 4 | 323 | 60 |