Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIG1
Entrez Gene ID
3978
Gene Name
DNA ligase 1
Gene Aliases
-
Chromosome Location
Chr.19: 48115445 - 48170603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000234.2 NP_000225.1 2 - 3 437 89
NM_001289064.1 NP_001275993.1 2 - 3 437 89
NM_001320970.1 NP_001307899.1 2 - 3 437 89
NR_110296.1 2 - 3 437 89
NR_135497.1 2 - 3 437 89
XM_017026809.1 XP_016882298.1 1 - 2 74 89
GenBank mRNA AB208791.1 - 2 - 3 160 89
AK225863.1 - 2 - 3 176 89
AK297296.1 - 2 - 3 148 89
AK303644.1 - 2 - 3 156 89
AK314210.1 - 2 - 3 144 89
BC108318.1 - 2 - 3 437 89
BC110622.2 - 1 - 2 86 89
M36067.1 - 2 - 3 137 89

View Full Product Details