The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004360.3 | NP_001004360.2 | 10 - 11 | 1050 | 112 |
NM_001178034.1 | NP_001171505.1 | 10 - 11 | 977 | 112 | |
NM_001178036.1 | NP_001171507.1 | 11 - 12 | 1454 | 112 | |
NM_001178037.1 | NP_001171508.1 | 10 - 11 | 1020 | 112 | |
NM_001321905.1 | NP_001308834.1 | 11 - 12 | 1480 | 112 | |
NM_001321906.1 | NP_001308835.1 | 11 - 12 | 1096 | 112 | |
NM_001321907.1 | NP_001308836.1 | 10 - 11 | 1365 | 112 | |
NM_001321908.1 | NP_001308837.1 | 10 - 11 | 1339 | 112 | |
NM_001321909.1 | NP_001308838.1 | 9 - 10 | 954 | 112 | |
NM_001321910.1 | NP_001308839.1 | 11 - 12 | 1423 | 112 | |
NM_001321911.1 | NP_001308840.1 | 10 - 11 | 1422 | 112 | |
NM_001321912.1 | NP_001308841.1 | 11 - 12 | 1454 | 112 | |
NM_001321913.1 | NP_001308842.1 | 9 - 10 | 1250 | 112 | |
NM_001321914.1 | NP_001308843.1 | 10 - 11 | 975 | 112 | |
NM_020868.4 | NP_065919.2 | 10 - 11 | 1365 | 112 | |
XM_017004565.1 | XP_016860054.1 | 11 - 12 | 1524 | 112 | |
XM_017004566.1 | XP_016860055.1 | 9 - 10 | 969 | 112 | |
GenBank mRNA | AB040925.2 | - | 10 - 11 | 932 | 112 |
AK289967.1 | - | 10 - 11 | 1064 | 112 | |
AK296490.1 | - | 10 - 11 | 1052 | 112 | |
AK315147.1 | - | 10 - 11 | 1169 | 112 | |
AY172661.3 | - | 10 - 11 | 944 | 112 | |
AY387785.1 | - | 10 - 11 | 966 | 112 | |
BC030832.1 | - | 10 - 11 | 1401 | 112 | |
DQ857321.1 | - | 10 - 11 | 932 | 112 | |
DQ857322.1 | - | 10 - 11 | 956 | 112 | |
DQ857323.1 | - | 10 - 11 | 923 | 112 |