The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004360.3 | NP_001004360.2 | 2 - 3 | 286 | 113 |
NM_001178034.1 | NP_001171505.1 | 2 - 3 | 213 | 113 | |
NM_001178036.1 | NP_001171507.1 | 3 - 4 | 690 | 113 | |
NM_001178037.1 | NP_001171508.1 | 2 - 3 | 256 | 113 | |
NM_001321905.1 | NP_001308834.1 | 3 - 4 | 716 | 113 | |
NM_001321906.1 | NP_001308835.1 | 3 - 4 | 332 | 113 | |
NM_001321907.1 | NP_001308836.1 | 2 - 3 | 601 | 113 | |
NM_001321910.1 | NP_001308839.1 | 3 - 4 | 659 | 113 | |
NM_001321911.1 | NP_001308840.1 | 2 - 3 | 658 | 113 | |
NM_001321912.1 | NP_001308841.1 | 3 - 4 | 690 | 113 | |
NM_001321914.1 | NP_001308843.1 | 2 - 3 | 216 | 113 | |
NM_020868.4 | NP_065919.2 | 2 - 3 | 601 | 113 | |
XM_017004565.1 | XP_016860054.1 | 3 - 4 | 760 | 113 | |
GenBank mRNA | AB040925.2 | - | 2 - 3 | 168 | 113 |
AK055256.1 | - | 4 - 5 | 774 | 113 | |
AK289967.1 | - | 2 - 3 | 300 | 113 | |
AK296490.1 | - | 2 - 3 | 288 | 113 | |
AK315147.1 | - | 2 - 3 | 405 | 113 | |
AY172661.3 | - | 2 - 3 | 180 | 113 | |
AY387785.1 | - | 2 - 3 | 202 | 113 | |
BC030832.1 | - | 2 - 3 | 637 | 113 | |
BI597037.1 | - | 2 - 3 | 243 | 113 | |
DQ857321.1 | - | 2 - 3 | 168 | 113 | |
DQ857322.1 | - | 2 - 3 | 192 | 113 | |
DQ857323.1 | - | 2 - 3 | 159 | 113 |