The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001896.2 | NP_001887.1 | 1 - 2 | 247 | 81 |
XM_005255800.3 | XP_005255857.1 | 1 - 2 | 485 | 81 | |
XM_005255801.3 | XP_005255858.1 | 1 - 2 | 247 | 81 | |
GenBank mRNA | AI150700.1 | - | 1 - 2 | 247 | 81 |
BC008812.2 | - | 1 - 2 | 121 | 81 | |
M55268.1 | - | 2 - 3 | 268 | 81 |