The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001896.2 | NP_001887.1 | 2 - 3 | 352 | 93 |
GenBank mRNA | AI150700.1 | - | 2 - 3 | 352 | 93 |
BC008812.2 | - | 2 - 3 | 226 | 93 | |
M55268.1 | - | 3 - 4 | 373 | 93 |