The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195226.1 | NP_001182155.1 | 3 - 4 | 303 | 83 |
NM_016335.4 | NP_057419.4 | 4 - 5 | 718 | 83 | |
GenBank mRNA | AB209472.1 | - | 3 - 4 | 369 | 83 |
BC036534.1 | - | 3 - 4 | 527 | 83 | |
BC068260.1 | - | 3 - 4 | 519 | 83 | |
BC094736.1 | - | 3 - 4 | 348 | 83 | |
BC118597.1 | - | 3 - 4 | 303 | 83 | |
BC121809.1 | - | 3 - 4 | 303 | 83 | |
KJ534920.1 | - | 3 - 4 | 514 | 83 | |
U79754.1 | - | 4 - 5 | 718 | 83 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006724935.3 | XP_006724998.1 | 3 - 4 | 547 | 83 |
XM_006724936.3 | XP_006724999.1 | 3 - 4 | 522 | 83 | |
XM_017030166.1 | XP_016885655.1 | 3 - 4 | 644 | 83 | |
XM_017030168.1 | XP_016885657.1 | 3 - 4 | 365 | 83 |