Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
Chromosome Location
Chr.22: 19130279 - 19144726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022719.2 NP_073210.1 2 - 3 342 117
NR_134304.1 2 - 3 342 117
XM_005261282.4 XP_005261339.1 2 - 3 347 117
XM_006724329.3 XP_006724392.1 2 - 3 706 117
XM_006724330.3 XP_006724393.1 2 - 3 747 117
XM_006724331.3 XP_006724394.1 2 - 3 747 117
XM_011530403.1 XP_011528705.1 2 - 3 391 117
XM_011530404.2 XP_011528706.1 2 - 3 350 117
XM_017028958.1 XP_016884447.1 2 - 3 706 117
GenBank mRNA BC003015.1 - 2 - 3 304 117
BC006542.2 - 2 - 3 306 117
BM545545.1 - 2 - 3 368 117
BX419278.2 - 2 - 3 350 117
CR456344.1 - 2 - 3 314 117
L77566.1 - 2 - 3 315 117
L78010.1 - 2 - 3 311 117

View Full Product Details