Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNDIG1
Entrez Gene ID
79953
Gene Name
synapse differentiation inducing 1
Gene Aliases
C20orf39, DSPC2, IFITMD5, TMEM90B
Chromosome Location
Chr.20: 24469199 - 24666617 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323606.1 NP_001310535.1 4 - 5 2141 72
NM_001323607.1 NP_001310536.1 3 - 4 817 72
NM_024893.2 NP_079169.1 3 - 4 1252 72
XM_017028065.1 XP_016883554.1 4 - 5 1613 72
XM_017028066.1 XP_016883555.1 3 - 4 970 72
XM_017028067.1 XP_016883556.1 3 - 4 876 72
XM_017028068.1 XP_016883557.1 3 - 4 1837 72
GenBank mRNA AK024282.1 - 3 - 4 1252 72
BC030637.1 - 3 - 4 754 72
CR457325.1 - 2 - 3 619 72

View Full Product Details